Tisk…

4-(Trimethylsilyloxy)benzaldehyde 97%

BLDPBD12239-1GEA 200 CZK
BLDPBD12239-1G BLDPBD12239-100G BLDPBD12239-5G BLDPBD12239-10G BLDPBD12239-25G
4-(Trimethylsilyloxy)benzaldehyde 97%
4-(Trimethylsilyloxy)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₁₄O₂Si
MW: 194,3025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040723
CAS číslo: 1012-12-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR