Tisk…

1,1-Bis(hydroxymethyl)cyclopentane 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD122925-250MGEA 1230 CZK
BLDPBD122925-250MG BLDPBD122925-5G BLDPBD122925-1G
1,1-Bis(hydroxymethyl)cyclopentane 98%
1,1-Bis(hydroxymethyl)cyclopentane

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₁₄O₂
MW: 130,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463707
CAS číslo: 5763-53-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR