Tisk…

Boc-D-cyclopropylglycine 97%

BLDPBD12354-5GEA 7670 CZK
BLDPBD12354-5G BLDPBD12354-25G BLDPBD12354-1G BLDPBD12354-250MG
Boc-D-cyclopropylglycine 97%
Boc-D-cyclopropylglycine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₇NO₄
MW: 215,246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659124
CAS číslo: 609768-49-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR