Tisk…

(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD12390-250MGEA 171 CZK
BLDPBD12390-250MG BLDPBD12390-5G BLDPBD12390-1G BLDPBD12390-100G BLDPBD12390-25G BLDPBD12390-10G
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 97%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₉NO₄
MW: 217,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094386
CAS číslo: 61478-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR