Chromone-3-carboxaldehyde 97%
|
|
BLDPBD12555-25GEA
1660
CZK
BLDPBD12555-25G
BLDPBD12555-10G
BLDPBD12555-1G
BLDPBD12555-100G
BLDPBD12555-5G
Chromone-3-carboxaldehyde 97%
Chromone-3-carboxaldehyde
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₁₀H₆O₃ MW: 174,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014667 CAS číslo: 17422-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...