4-Chloro-8-methoxyquinoline 98%
|
|
BLDPBD12687-250MGEA
428
CZK
BLDPBD12687-250MG
BLDPBD12687-1G
BLDPBD12687-5G
4-Chloro-8-methoxyquinoline 98%
4-Chloro-8-methoxyquinoline
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₁₀H₈ClNO MW: 193,6296 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00033679 CAS číslo: 16778-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...