6-Methoxy-8-nitroquinoline 98%
|
|
BLDPBD12732-10GEA
1430
CZK
BLDPBD12732-10G
BLDPBD12732-100G
BLDPBD12732-5G
BLDPBD12732-1G
BLDPBD12732-25G
6-Methoxy-8-nitroquinoline 98%
6-Methoxy-8-nitroquinoline
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₈N₂O₃ MW: 204,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006802 CAS číslo: 85-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...