Tisk…

Ethyl-2-bromothiazole-4-carboxylate 97%

BLDPBD128225-1GEA 200 CZK
BLDPBD128225-1G BLDPBD128225-25G BLDPBD128225-5G BLDPBD128225-250MG BLDPBD128225-500G BLDPBD128225-10G BLDPBD128225-100G
Ethyl-2-bromothiazole-4-carboxylate 97%
Ethyl-2-bromothiazole-4-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₆BrNO₂S
MW: 236,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788564
CAS číslo: 100367-77-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR