Tisk…

Ethyl-3,5-bis(trifluoromethyl)phenylacetate 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD128541-5GEA 2090 CZK
BLDPBD128541-5G BLDPBD128541-25G BLDPBD128541-1G
Ethyl-3,5-bis(trifluoromethyl)phenylacetate 97%
Ethyl-3,5-bis(trifluoromethyl)phenylacetate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₀F₆O₂
MW: 300,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689894
CAS číslo: 144632-97-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR