Tisk…

Dimethyl-3-nitrophthalate 99%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD12908-25GEA 6960 CZK
BLDPBD12908-25G BLDPBD12908-1G BLDPBD12908-100G BLDPBD12908-5G
Dimethyl-3-nitrophthalate 99%
Dimethyl-3-nitrophthalate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₉NO₆
MW: 239,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017184
CAS číslo: 13365-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR