Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD129502-1GEA
1140
CZK
BLDPBD129502-1G
BLDPBD129502-250MG
BLDPBD129502-5G
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 98%
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
Storage condition: Sealed in dry, 2-8°C
Vzorec:
C₁₀H₁₀N₂O₂ MW: 190,1986 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06411542 CAS číslo: 16205-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...