Tisk…

N,N-Dimethyl-1-dodecylamine N-oxide 98+%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD136053-1GEA 912 CZK
BLDPBD136053-1G BLDPBD136053-5G BLDPBD136053-25G
N,N-Dimethyl-1-dodecylamine N-oxide 98+%
N,N-Dimethyl-1-dodecylamine N-oxide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₄H₃₁NO
MW: 229,40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002049
CAS číslo: 1643-20-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR