Tisk…

3-(Thien-3-yl)benzaldehyde 95%

BLDPBD13669-100MGEA 1860 CZK
BLDPBD13669-100MG BLDPBD13669-1G BLDPBD13669-250MG
3-(Thien-3-yl)benzaldehyde 95%
3-(Thien-3-yl)benzaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₁H₈OS
MW: 188,246 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039150
CAS číslo: 129746-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR