Tisk…

N-(4-Methoxybenzylidene)-4-butylaniline 98%

BLDPBD137257-25GEA 3880 CZK
BLDPBD137257-25G BLDPBD137257-1G BLDPBD137257-5G
N-(4-Methoxybenzylidene)-4-butylaniline 98%
N-(4-Methoxybenzylidene)-4-butylaniline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₈H₂₁NO
MW: 267,3654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009464
CAS číslo: 26227-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR