Tisk…

Methyl-3-cyano-5-fluorobenzoate 95+%

BLDPBD138646-250MGEA 827 CZK
BLDPBD138646-250MG BLDPBD138646-1G
Methyl-3-cyano-5-fluorobenzoate 95+%
Methyl-3-cyano-5-fluorobenzoate
Vzorec: C₉H₆FNO₂
MW: 179,15 g/mol
MDL Number: MFCD08688582
CAS číslo: 886732-29-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR