Tisk…

Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate 98+%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD138918-250MGEA 2310 CZK
BLDPBD138918-250MG BLDPBD138918-1G BLDPBD138918-100MG
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate 98+%
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₁₁BrN₂O₂
MW: 247,0891 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00173775
CAS číslo: 175276-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR