Tisk…

Methyl-4-fluorosalicylate 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD138979-10GEA 3820 CZK
BLDPBD138979-10G BLDPBD138979-5G BLDPBD138979-25G BLDPBD138979-1G BLDPBD138979-250MG
Methyl-4-fluorosalicylate 98%
Methyl-4-fluorosalicylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₇FO₃
MW: 170,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093965
CAS číslo: 392-04-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR