Tisk…

Methyl 2-bromo-4-(bromomethyl)benzoate 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD139775-250MGEA 542 CZK
BLDPBD139775-250MG BLDPBD139775-25G BLDPBD139775-100MG BLDPBD139775-1G BLDPBD139775-10G BLDPBD139775-5G
Methyl 2-bromo-4-(bromomethyl)benzoate 95%
Methyl 2-bromo-4-(bromomethyl)benzoate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₈Br₂O₂
MW: 307,967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03840738
CAS číslo: 128577-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR