1-Benzyl-4-hydroxypiperidine 98%
|
|
BLDPBD14189-10GEA
285
CZK
BLDPBD14189-10G
BLDPBD14189-500G
BLDPBD14189-0.5G
BLDPBD14189-5G
BLDPBD14189-100G
BLDPBD14189-25G
1-Benzyl-4-hydroxypiperidine 98%
1-Benzyl-4-hydroxypiperidine
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₁₂H₁₇NO MW: 191,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006503 CAS číslo: 4727-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...