Tisk…

N,N-Dimethylcyanoacetamide 98%

BLDPBD142210-25GEA 6130 CZK
BLDPBD142210-25G BLDPBD142210-1G BLDPBD142210-5G
N,N-Dimethylcyanoacetamide 98%
N,N-Dimethylcyanoacetamide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₅H₈N₂O
MW: 112,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00051623
CAS číslo: 7391-40-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR