Tisk…

2-Amino-4-(1-naphthyl)thiazole 98%

BLDPBD14526-5GEA 5330 CZK
BLDPBD14526-5G BLDPBD14526-1G BLDPBD14526-25G BLDPBD14526-250MG
2-Amino-4-(1-naphthyl)thiazole 98%
2-Amino-4-(1-naphthyl)thiazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₃H₁₀N₂S
MW: 226,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236016
CAS číslo: 56503-96-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR