Tisk…

Potassium allyltrifluoroborate 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD148727-100GEA 11860 CZK
BLDPBD148727-100G BLDPBD148727-25G BLDPBD148727-500G BLDPBD148727-10G BLDPBD148727-5G BLDPBD148727-250MG BLDPBD148727-1G
Potassium allyltrifluoroborate 95%
Potassium allyltrifluoroborate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₃H₅BF₃K
MW: 147,976 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368222
CAS číslo: 233664-53-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR