Tisk…

Potassium benzyltrifluoroborate 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD148736-100GEA 37570 CZK
BLDPBD148736-100G BLDPBD148736-10G BLDPBD148736-250MG BLDPBD148736-5G BLDPBD148736-25G BLDPBD148736-1G
Potassium benzyltrifluoroborate 97%
Potassium benzyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₇BF₃K
MW: 198,0350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427261
CAS číslo: 329976-73-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR