Tisk…

Potassium 2-phenylethyltrifluoroborate 97%

BLDPBD148868-250MGEA 684 CZK
BLDPBD148868-250MG BLDPBD148868-5G BLDPBD148868-1G BLDPBD148868-25G
Potassium 2-phenylethyltrifluoroborate 97%
Potassium 2-phenylethyltrifluoroborate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₈H₉BF₃K
MW: 212,06 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09039257
CAS číslo: 329976-74-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR