Propargyl benzenesulphonate 97%
|
|
BLDPBD149205-100GEA
1570
CZK
BLDPBD149205-100G
BLDPBD149205-0.025
BLDPBD149205-10G
BLDPBD149205-5G
BLDPBD149205-25G
Propargyl benzenesulphonate 97%
Propargyl benzenesulphonate
Storage condition: Sealed in dry, 2-8°C
Vzorec:
C₉H₈O₃S MW: 196,2200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00013459 CAS číslo: 6165-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...