Tisk…

Propargyl-p-toluenesulfonate 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD149213-5GEA 1060 CZK
BLDPBD149213-5G BLDPBD149213-1G BLDPBD149213-100G BLDPBD149213-25G
Propargyl-p-toluenesulfonate 95%
Propargyl-p-toluenesulfonate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₁₀O₃S
MW: 210,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01462194
CAS číslo: 6165-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR