Tisk…

4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 98%

BLDPBD14944-250MGEA 741 CZK
BLDPBD14944-250MG BLDPBD14944-1G BLDPBD14944-5G BLDPBD14944-25G
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 98%
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₃H₁₉BO₄
MW: 250,099 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02093723
CAS číslo: 269410-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR