2-Pyridinesulphonic acid 98%
|
|
BLDPBD149747-250MGEA
285
CZK
BLDPBD149747-250MG
BLDPBD149747-1G
BLDPBD149747-10G
BLDPBD149747-5G
BLDPBD149747-25G
2-Pyridinesulphonic acid 98%
2-Pyridinesulphonic acid
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₅H₅NO₃S MW: 159,1631 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234080 CAS číslo: 15103-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...