Tisk…

7-Chloropyrido[3,4-b]pyrazine 95%

BLDPBD149790-250MGEA 1430 CZK
BLDPBD149790-250MG BLDPBD149790-1G BLDPBD149790-100MG BLDPBD149790-5G
7-Chloropyrido[3,4-b]pyrazine 95%
7-Chloropyrido[3,4-b]pyrazine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₄ClN₃
MW: 165,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044722
CAS číslo: 93049-39-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR