Tisk…

2'-Chloro-4'-(4-chlorophenoxy)acetophenone 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD15148-25GEA 941 CZK
BLDPBD15148-25G BLDPBD15148-100G BLDPBD15148-5G
2'-Chloro-4'-(4-chlorophenoxy)acetophenone 97%
2'-Chloro-4'-(4-chlorophenoxy)acetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₄H₁₀Cl₂O₂
MW: 281,1340 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140226
CAS číslo: 119851-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR