Tetrabutylammonium toluene-4-sulphonate 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD153271-5GEA
257
CZK
BLDPBD153271-5G
BLDPBD153271-25G
BLDPBD153271-100G
BLDPBD153271-500G
Tetrabutylammonium toluene-4-sulphonate 98%
Tetrabutylammonium toluene-4-sulphonate
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₂₃H₄₃NO₃S MW: 413,6574 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043227 CAS číslo: 7182-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...