Tisk…

Thioacetanilide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD153880-5GEA 2710 CZK
BLDPBD153880-5G BLDPBD153880-1G BLDPBD153880-250MG
Thioacetanilide 98%
Thioacetanilide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₉NS
MW: 151,2288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004942
CAS číslo: 637-53-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR