Trithiocyanuric acid 98%
|
|
BLDPBD155962-100GEA
485
CZK
BLDPBD155962-100G
BLDPBD155962-500G
BLDPBD155962-10G
BLDPBD155962-1000G
BLDPBD155962-25G
Trithiocyanuric acid 98%
Trithiocyanuric acid
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₃H₃N₃S₃ MW: 177,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006052 CAS číslo: 638-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...