1,4-Anthraquinone 98%
BLDPBD15686-5GEA
513
CZK
BLDPBD15686-5G
BLDPBD15686-25G
BLDPBD15686-1G
1,4-Anthraquinone 98%
1,4-Anthraquinone
Vzorec:
C₁₄H₈O₂ MW: 208,22 g/mol |
MDL Number:
MFCD00013724 CAS číslo: 635-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...