(1-Ethoxycarbonylethylidene)triphenylphosphorane 97% (by HPLC)
|
|
BLDPBD1576-10GEA
285
CZK
BLDPBD1576-10G
BLDPBD1576-1000G
BLDPBD1576-25G
BLDPBD1576-5G
BLDPBD1576-500G
BLDPBD1576-100G
(1-Ethoxycarbonylethylidene)triphenylphosphorane 97% (by HPLC)
(1-Ethoxycarbonylethylidene)triphenylphosphorane
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Vzorec:
C₂₃H₂₃O₂P MW: 362,41 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00009160 CAS číslo: 5717-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...