Tisk…

2-Hydroxythiazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD157703-1GEA 2310 CZK
BLDPBD157703-1G BLDPBD157703-25G BLDPBD157703-5G BLDPBD157703-10G BLDPBD157703-250MG
2-Hydroxythiazole 97%
2-Hydroxythiazole

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₃H₃NOS
MW: 101,1270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094166
CAS číslo: 82358-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR