Tisk…

1-Boc-4-chlorosulphonylpiperidine 98%

BLDPBD157812-25GEA 25630 CZK
BLDPBD157812-25G BLDPBD157812-10G BLDPBD157812-250MG BLDPBD157812-5G BLDPBD157812-100MG BLDPBD157812-1G
1-Boc-4-chlorosulphonylpiperidine 98%
1-Boc-4-chlorosulphonylpiperidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₁₈ClNO₄S
MW: 283,767 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01861750
CAS číslo: 782501-25-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR