5-Iodo-1-methyl-1H-pyrazole 97%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD157840-25GEA
8070
CZK
BLDPBD157840-25G
BLDPBD157840-5G
BLDPBD157840-10G
BLDPBD157840-250MG
BLDPBD157840-1G
BLDPBD157840-100G
5-Iodo-1-methyl-1H-pyrazole 97%
5-Iodo-1-methyl-1H-pyrazole
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₄H₅IN₂ MW: 208,00 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11109327 CAS číslo: 34091-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...