Tisk…

Methyl-3-amino-5-chlorobenzoate 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD159396-250MGEA 684 CZK
BLDPBD159396-250MG
Methyl-3-amino-5-chlorobenzoate 98%
Methyl-3-amino-5-chlorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₈ClNO₂
MW: 185,6076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053764
CAS číslo: 21961-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR