Dibenzyl carbonate 98%
|
|
BLDPBD15941-25GEA
8640
CZK
BLDPBD15941-25G
BLDPBD15941-5G
BLDPBD15941-10G
BLDPBD15941-1G
Dibenzyl carbonate 98%
Dibenzyl carbonate
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₁₅H₁₄O₃ MW: 242,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014436 CAS číslo: 3459-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...