3-Bromo-2-methylanisole 97%
|
|
BLDPBD160072-1GEA
456
CZK
BLDPBD160072-1G
BLDPBD160072-25G
BLDPBD160072-100G
BLDPBD160072-0.025
BLDPBD160072-5G
BLDPBD160072-10G
3-Bromo-2-methylanisole 97%
3-Bromo-2-methylanisole
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₈H₉BrO MW: 201,0605 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100991 CAS číslo: 31804-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...