Tisk…

4,6-Dichloro-2-(trifluoromethyl)pyrimidine 98%

BLDPBD160306-10GEA 6790 CZK
BLDPBD160306-10G BLDPBD160306-5G BLDPBD160306-100MG BLDPBD160306-1G BLDPBD160306-250MG BLDPBD160306-25G
4,6-Dichloro-2-(trifluoromethyl)pyrimidine 98%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₅HCl₂F₃N₂
MW: 216,98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436597
CAS číslo: 705-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR