Tisk…

2-Bromo-3-chloroaniline 98%

BLDPBD161241-500GEA 57860 CZK
BLDPBD161241-0.5G BLDPBD161241-500G BLDPBD161241-5G BLDPBD161241-250MG BLDPBD161241-1G BLDPBD161241-100G BLDPBD161241-25G BLDPBD161241-10G
2-Bromo-3-chloroaniline 98%
2-Bromo-3-chloroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₅BrClN
MW: 206,4700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699543
CAS číslo: 96558-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR