Tisk…

2,4-Dichloro-6-fluorobenzoic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD162122-5GEA 7760 CZK
BLDPBD162122-5G BLDPBD162122-250MG BLDPBD162122-1G BLDPBD162122-10G
2,4-Dichloro-6-fluorobenzoic acid 95%
2,4-Dichloro-6-fluorobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₃Cl₂FO₂
MW: 209,0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12404936
CAS číslo: 904285-09-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR