Tisk…

2-Amino-4-hydroxybenzoic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD162450-100MGEA 2340 CZK
BLDPBD162450-100MG BLDPBD162450-250MG
2-Amino-4-hydroxybenzoic acid 97%
2-Amino-4-hydroxybenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₇NO₃
MW: 153,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071506
CAS číslo: 38160-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR