6-Bromo-3-iodo-1H-indazole 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD162592-25GEA
6470
CZK
BLDPBD162592-25G
BLDPBD162592-100G
BLDPBD162592-1G
BLDPBD162592-5G
6-Bromo-3-iodo-1H-indazole 98%
6-Bromo-3-iodo-1H-indazole
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₇H₄BrIN₂ MW: 322,9285 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07781599 CAS číslo: 885521-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...