Tisk…

1-(4-Bromophenyl)-cyclobutanemethanamine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD162768-100MGEA 2800 CZK
BLDPBD162768-100MG BLDPBD162768-250MG BLDPBD162768-1G
1-(4-Bromophenyl)-cyclobutanemethanamine 98%
1-(4-Bromophenyl)-cyclobutanemethanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₁H₁₄BrN
MW: 240,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11188929
CAS číslo: 1039932-36-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR