Tisk…

1,4-Bis(methylamino)-9,10-anthraquinone 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD16301-1GEA 171 CZK
BLDPBD16301-1G BLDPBD16301-5G
1,4-Bis(methylamino)-9,10-anthraquinone 97%
1,4-Bis(methylamino)-9,10-anthraquinone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₆H₁₄N₂O₂
MW: 266,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001198
CAS číslo: 2475-44-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR