(4-Benzamidophenyl)boronic acid 97%
|
|
BLDPBD163780-1GEA
1830
CZK
BLDPBD163780-1G
BLDPBD163780-5G
BLDPBD163780-250MG
(4-Benzamidophenyl)boronic acid 97%
(4-Benzamidophenyl)boronic acid
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₁₃H₁₂BNO₃ MW: 241,05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08235089 CAS číslo: 397843-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...