Tisk…

Fmoc-D-phenylalanine 98%

BLDPBD1646-100GEA 2090 CZK
BLDPBD1646-100G BLDPBD1646-10G BLDPBD1646-500G BLDPBD1646-5G BLDPBD1646-25G BLDPBD1646-0.5G
Fmoc-D-phenylalanine 98%
Fmoc-D-phenylalanine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₂₄H₂₁NO₄
MW: 387,4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00062955
CAS číslo: 86123-10-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR